missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium slug, 9.5mm (0.37 in.) dia. x 9.5mm (0.37 in.) length, 99.9+% (metals basis)

Manufacturer:  Alfa Aesar™ 10381QI

 View more versions of this product

Catalog No. AA10381QI



Specifications

Specifications

Titanium slug
1 slug
7440-32-6
Ti
alloy
[Ti]
Odorless
23963
Slug
≈3g/slug
9.5mm (0.37 in.) dia. x 9.5mm (0.37 in.) length
(metals basis)
MFCD00011264
RTAQQCXQSZGOHL-UHFFFAOYSA-N
titanium
47.867
CHEBI:33341
≥99.9%

Chemical Identifiers

7440-32-6
47.867
RTAQQCXQSZGOHL-UHFFFAOYSA-N
23963
titanium
Ti
MFCD00011264
alloy
CHEBI:33341
[Ti]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents