Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium slug, 9.5mm (0.37 in.) dia. x 9.5mm (0.37 in.) length, 99.9+% (metals basis)

Chemical Identifiers

Chemical Name or Material Titanium slug
CAS 7440-32-6
Name Note 9.5mm (0.37 in.) dia. x 9.5mm (0.37 in.) length
MDL Number MFCD00011264
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA10381QI

 
alfa aesar™
10381QI
1 slug CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA10381QJ

 
alfa aesar™
10381QJ
5 slug CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA10381QK

 
alfa aesar™
10381QK
25 slug CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Titanium slug
TSCA TSCA
Name Note 9.5mm (0.37 in.) dia. x 9.5mm (0.37 in.) length
Physical Form Slug
Assay ≥99.9%
Assay Percent Notes (metals basis)
MDL Number MFCD00011264
CAS 7440-32-6
Odor Odorless
Weight ≈3g/slug
Quantity 1 slug