Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium silicide sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.5% (metals basis)

Chemical Identifiers

Chemical Name or Material Titanium silicide sputtering target
Name Note 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick
Formula Weight 104.07
MDL Number MFCD00151541
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41116KS

  SDS
alfa aesar™
41116KS
1 each CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 104.07
Assay 99.5%
Assay Percent Notes (metals basis)
MDL Number MFCD00151541
Quantity 1 each
Chemical Name or Material Titanium silicide sputtering target
TSCA TSCA
Name Note 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick