Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium silicide, 99.5% (metals basis)

$130.64 - $130.64

Chemical Identifiers

Chemical Name or Material Titanium silicide
CAS 12039-83-7
Linear Formula TiSi2
Formula Weight 104.07
MDL Number MFCD00151541
UN Number UN3178
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1308818

  SDS
alfa aesar™
1308818
50g Each for $130.64

AA1308830

  SDS
alfa aesar™
1308830
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 104.07
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 12039-83-7
Color Gray
Assay Percent Notes (metals basis)
Health Hazard 2 GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 99.5%
Melting Point 1760°C
Quantity 50g
DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
Linear Formula TiSi2
MDL Number MFCD00151541
Chemical Name or Material Titanium silicide
Health Hazard 1 Warning
TSCA TSCA
RTECS Number XR2420000
Mesh Size −200 Mesh
Physical Form Powder
UN Number UN3178
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.