missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Titanium Silicide, 99.5% (Metal Basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1308730

 View more versions of this product

Catalog No. AA1308730



Specifications

Specifications

Titanium Silicide
99.5%, metal basis
MFCD01310208
GNKTZDSRQHMHLZ-UHFFFAOYSA-N
silicon;titanium
6336553
-325 Mesh Powder
12067-57-1
Si3Ti5
silicon; titanium, titanium silicide ti5si3, pentatitanium trisilicide, titanium silicide,-325 mesh
[Si].[Si].[Si].[Ti].[Ti].[Ti].[Ti].[Ti]
323.59
323.76
250g

Chemical Identifiers

12067-57-1
323.59
GNKTZDSRQHMHLZ-UHFFFAOYSA-N
6336553
[Si].[Si].[Si].[Ti].[Ti].[Ti].[Ti].[Ti]
Si3Ti5
MFCD01310208
silicon; titanium, titanium silicide ti5si3, pentatitanium trisilicide, titanium silicide,-325 mesh
silicon;titanium
Safety and Handling

Safety and Handling

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves. This material and/or its container must be disposed of as hazardous waste.

Flammable. Irritating to eyes, respiratory system and skin.

DOTInformation : DOT Class: 4.1

EINECSNumber : 235-082-9

Documents