missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Titanium rod, 12.7mm (0.5in) dia, annealed, 99.5% (metals basis), Thermo Scientific Chemicals
Form: approx. 5.7g/cm
Supplier: Thermo Scientific Chemicals 010394CB
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Silver to Gray | |
Annealed | |
Silver Gray rod | |
Ti | |
250 cm | |
RTAQQCXQSZGOHL-UHFFFAOYSA-N | |
titanium | |
23963 | |
Odorless |
Titanium | |
7440-32-6 | |
99.5% (Metals basis) | |
MFCD00011264,MFCD00151301 | |
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1 | |
[Ti] | |
47.87 | |
CHEBI:33341 | |
Rod |
Chemical Identifiers
7440-32-6 | |
47.87 | |
RTAQQCXQSZGOHL-UHFFFAOYSA-N | |
23963 | |
titanium |
Ti | |
MFCD00011264,MFCD00151301 | |
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1 | |
CHEBI:33341 | |
[Ti] |
Safety and Handling
EINECSNumber : 231-142-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only