missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Titanium rod, 99.5% (metals basis), Alfa Aesar™

Form: approx. 5.7g/cm

Manufacturer:  Thermo Fisher Scientific 10394BS

 View more versions of this product

Catalog No. AA10394BS



Specifications

Specifications

Titanium
Annealed
Silver Gray rod
Ti
alloy
[Ti]
Odorless
23963
Rod, 12.7mm (0.5 in.) dia.
10cm
7440-32-6
99.5% (Metals basis)
MFCD00011264
RTAQQCXQSZGOHL-UHFFFAOYSA-N
titanium
47.867
CHEBI:33341
Silver to Gray

Chemical Identifiers

7440-32-6
47.867
RTAQQCXQSZGOHL-UHFFFAOYSA-N
23963
titanium
Ti
MFCD00011264
alloy
CHEBI:33341
[Ti]
Safety and Handling

Safety and Handling

EINECSNumber : 231-142-3

Documents