missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Titanium Rod, 6.4mm (0.25 in.) dia., 99.99% (Metals basis), Alfa Aesar™

Form: Approx. 1.45g/cm

Manufacturer:  Alfa Aesar 14001CZ

 View more versions of this product

Catalog No. AA14001CZ



Specifications

Specifications

Titanium
7440-32-6
99.99% (Metals basis)
MFCD00011264
RTAQQCXQSZGOHL-UHFFFAOYSA-N
titanium
47.867
CHEBI:33341
Silver to Gray
4 x 25cm
Silver Gray rod
Ti
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
[Ti]
Odorless
23963
Rod, 6.4mm (0.25 in.) dia.

Chemical Identifiers

7440-32-6
47.867
RTAQQCXQSZGOHL-UHFFFAOYSA-N
23963
titanium
Ti
MFCD00011264
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
CHEBI:33341
[Ti]
Documents