Please login to your online account to display your discounted pricing

Titanium, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical


Quantity 100mL
Packaging Poly Bottle

Manufacturer: fisher scientific company  FLST96100

 View more versions of this product

Catalog No. ST96-100

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Melting Point 0°C
Boiling Point 100°C
Purity Grade Certified Grade
Quantity 100mL
DOT Information DOT Class 8, : Corrosive
Color Red
Physical Form Liquid
Vapor Pressure 14mmHg at 20°C
Density 0.7g/cm³
Traceability to NIST LOT NUMBER USED TITANIUM STANDARD
CAS 7732-18-5
CAS 7647-01-0
ChemAlert Storage Symbol White
Chemical Name or Material Titanium, Reference Standard Solution
MDL Number MFCD00011264
CAS 7722-84-1
Concentration 990 to 1010ppm
Packaging Poly Bottle
Name Note 1000ppm ±1%
CAS 7440-32-6

Specifications

Chemical Name or Material Titanium, Reference Standard Solution
Name Note 1000ppm ±1%
CAS 7647-01-0
CAS 7440-32-6
CAS 7732-18-5
CAS 7722-84-1
Purity Grade Certified Grade
MDL Number MFCD00011264
Accessories
 

SDS & Safety Documents