Please login to your online account to display your discounted pricing

Titanium, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical

$103.69 - $103.69

Chemical Identifiers

Chemical Name or Material Titanium, Reference Standard Solution
Name Note 1000ppm ±1%
CAS 7647-01-0
CAS 7440-32-6
CAS 7732-18-5
CAS 7722-84-1
Purity Grade Certified Grade
MDL Number MFCD00011264
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

ST96-100

  SDS
fisher scientific company
FLST96100
Poly Bottle 100mL Each for $103.69
Case of 6 Each for $622.14
Description & Specifications

Specifications

Chemical Name or Material Titanium, Reference Standard Solution
Name Note 1000ppm ±1%
CAS 7647-01-0
ChemAlert Storage Symbol White
CAS 7440-32-6
CAS 7732-18-5
DOT Information DOT Class 8, : Corrosive
CAS 7722-84-1
Purity Grade Certified Grade
Physical Form Liquid
Concentration 990 to 1010ppm
MDL Number MFCD00011264
Packaging Poly Bottle
Traceability to NIST LOT NUMBER USED TITANIUM STANDARD
Vapor Pressure 14mmHg at 20°C
Density 0.7g/cm³
Melting Point 0°C
Boiling Point 100°C
Color Red
Quantity 100mL
Safety and Handling