missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium powder, -100 mesh, 99.4%

Manufacturer:  Alfa Aesar™ 1038314

 View more versions of this product

Catalog No. AA1038314



Specifications

Specifications

Titanium powder
7440-32-6
MFCD00011264
−100 Mesh
RTAQQCXQSZGOHL-UHFFFAOYSA-N
titanium
47.867
CHEBI:33341
99.4%
25g
Ti
UN2546
alloy
[Ti]
Odorless
23963
Powder
Gray

Chemical Identifiers

7440-32-6
47.867
RTAQQCXQSZGOHL-UHFFFAOYSA-N
23963
titanium
Ti
MFCD00011264
alloy
CHEBI:33341
[Ti]
Safety and Handling

Safety and Handling

GHS H Statement
H252
Self-heating in large quantities
may catch fire.

GHS P Statement
P280-P235+P410-P420-P407-P413a
Wear protective gloves/protective clothing/eye protection/face protection.
Keep cool.
Protect from sunlight.
Store away from other materials.
Maintain air gap between stacks/pallets.
Store bulk masses greater than 1 lb. at temperatures not exceeding 100°F.

Warning

DOTInformation : Transport Hazard Class: 4.2; Packing Group: III; Proper Shipping Name: TITANIUM POWDER, DRY

RTECSNumber : XR1700000

TSCA : TSCA

Documents