Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium plate, 6mm (0.2 in.) thick, 100x100mm (3.9x3.9 in.), Puratronic™, 99.999% (metals basis)

Chemical Identifiers

Chemical Name or Material Titanium plate
CAS 7440-32-6
Name Note 6mm (0.2 in.) thick, 100x100mm (3.9 x 3.9 in.)
Purity Grade Puratronic™
MDL Number MFCD00011264
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA36680KG

  SDS
alfa aesar™
36680KG
1 piece CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Titanium plate
TSCA TSCA
Name Note 6mm (0.2 in.) thick, 100x100mm (3.9 x 3.9 in.)
Purity Grade Puratronic™
Physical Form Plate
Assay 99.999%
Assay Percent Notes (metals basis)
MDL Number MFCD00011264
CAS 7440-32-6
Odor Odorless
Weight ≈271g/pc
Quantity 1 piece