missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium plate, 3.175mm (0.125 in.) thick, 100x100mm (3.9x3.9 in.), annealed, 99.5% (metals basis)

Manufacturer:  Alfa Aesar™ 12598KJ

 View more versions of this product

Catalog No. AA12598KJ



Specifications

Specifications

Titanium plate
5 pc.
7440-32-6
Ti
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
[Ti]
Odorless
23963
Plate
≈143g/pc
3.175mm (0.125 in.) thick, 100x100mm (3.9x3.9 in.), Annealed
(metals basis)
MFCD00011264
RTAQQCXQSZGOHL-UHFFFAOYSA-N
titanium
47.867
CHEBI:33341
99.5%

Chemical Identifiers

7440-32-6
47.867
RTAQQCXQSZGOHL-UHFFFAOYSA-N
23963
titanium
Ti
MFCD00011264
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
CHEBI:33341
[Ti]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents