missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium, plasma standard solution, Specpure™, Ti 1000μg/mL

Chemical Identifiers

MDL Number MFCD00011264
Chemical Name or Material Titanium plasma standard solution, Ti 1000μg/mL
Grade Specpure™
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA38720AD SDS Alfa Aesar™
38720AD
50mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA38720AE SDS Alfa Aesar™
38720AE
100mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA38720AP SDS Alfa Aesar™
38720AP
500mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

MFCD00011264
Titanium plasma standard solution, Ti 1000μg/mL
Specpure™

Specifications

Liquid
50mL
Specpure™
MFCD00011264
Titanium plasma standard solution, Ti 1000μg/mL

Safety and Handling

TSCA : TSCA