missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Titanium, plasma standard solution, Specpure™, Ti 10,000μg/ml, Alfa Aesar™

Chemical Identifiers

CAS 7440-32-6
Molecular Formula Ti
Molecular Weight (g/mol) 47.867
MDL Number MFCD00011264
InChI Key RTAQQCXQSZGOHL-UHFFFAOYSA-N
Synonym alloy, c.p., contimet 30, cp, ii hydride, jistp28, oremet, titan vt 1-1, titanium-125, vt1
PubChem CID 23963
ChEBI CHEBI:33341
IUPAC Name titanium
SMILES [Ti]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA38725AD
View Documents
Thermo Fisher Scientific
38725AD
50mL N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA38725AE
View Documents
Thermo Fisher Scientific
38725AE
100mL N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA38725AP
View Documents
Thermo Fisher Scientific
38725AP
500mL N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

7440-32-6
47.867
RTAQQCXQSZGOHL-UHFFFAOYSA-N
23963
titanium
Ti
MFCD00011264
alloy, c.p., contimet 30, cp, ii hydride, jistp28, oremet, titan vt 1-1, titanium-125, vt1
CHEBI:33341
[Ti]

Specifications

7440-32-6
MFCD00011264
RTAQQCXQSZGOHL-UHFFFAOYSA-N
titanium
23963
Liquid
50mL
Ti
alloy, c.p., contimet 30, cp, ii hydride, jistp28, oremet, titan vt 1-1, titanium-125, vt1
[Ti]
47.867
CHEBI:33341
Plastic bottle
Titanium
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures