missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Titanium, plasma standard solution, Specpure™, Ti 10,000μg/ml, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 38725AD

 View more versions of this product

Catalog No. AA38725AD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Titanium
Ti
alloy, c.p., contimet 30, cp, ii hydride, jistp28, oremet, titan vt 1-1, titanium-125, vt1
[Ti]
47.867
CHEBI:33341
Plastic bottle
7440-32-6
MFCD00011264
RTAQQCXQSZGOHL-UHFFFAOYSA-N
titanium
23963
Liquid
50mL

Chemical Identifiers

7440-32-6
47.867
RTAQQCXQSZGOHL-UHFFFAOYSA-N
23963
titanium
Ti
MFCD00011264
alloy, c.p., contimet 30, cp, ii hydride, jistp28, oremet, titan vt 1-1, titanium-125, vt1
CHEBI:33341
[Ti]
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents