Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium, Oil based standard solution, Specpure™, Ti 1000μg/g

Chemical Identifiers

Chemical Name or Material Titanium Oil based standard solution
Name Note Ti 1000μg/g
Purity Grade Specpure™
MDL Number MFCD00011264
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1420418

  SDS
alfa aesar™
1420418
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 50g
Name Note Ti 1000μg/g
MDL Number MFCD00011264
Purity Grade Specpure™
Chemical Name or Material Titanium Oil based standard solution
Physical Form Liquid