Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium nitride sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.5% (metals basis)

$1,330.47 - $1,330.47

Chemical Identifiers

Chemical Name or Material Titanium nitride sputtering target
CAS 25583-20-4
Name Note 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick
Formula Weight 61.91
MDL Number MFCD00049596
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41112KS

  SDS
alfa aesar™
41112KS
1 each Each for $1,330.47
Description & Specifications

Specifications

Formula Weight 61.91
Physical Form Hot Pressed Material
Name Note 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick
TSCA TSCA
Melting Point 2950°C
RTECS Number XR2230000
MDL Number MFCD00049596
CAS 25583-20-4
Assay Percent Notes (metals basis)
Quantity 1 each
Color Gray
Density 5.22g/mL
Assay 99.5%
Chemical Name or Material Titanium nitride sputtering target