Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium nitride sputtering target, 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.5% (metals basis)

$1,095.97 - $1,095.97

Chemical Identifiers

Chemical Name or Material Titanium nitride sputtering target
CAS 25583-20-4
Name Note 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
Formula Weight 61.91
MDL Number MFCD00049596
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41111KS

  SDS
alfa aesar™
41111KS
1 each Each for $1,095.97
Description & Specifications

Specifications

Physical Form Hot Pressed Material
Formula Weight 61.91
Chemical Name or Material Titanium nitride sputtering target
TSCA TSCA
Name Note 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
Assay 99.5%
Assay Percent Notes (metals basis)
MDL Number MFCD00049596
CAS 25583-20-4
Color Gray
Melting Point 2950°C
Quantity 1 each
Density 5.22g/mL
RTECS Number XR2230000