missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium nitride sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.5% (metals basis)

Magnetron sputtering source

$936.45 - $936.45

Chemical Identifiers

CAS 25583-20-4
Formula Weight 61.91
MDL Number MFCD00049596
Chemical Name or Material Titanium nitride sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA41110KS SDS Alfa Aesar™
41110KS
1 each Each for $936.45
Specifications Description & Specifications

Chemical Identifiers

25583-20-4
61.91
MFCD00049596
Titanium nitride sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick

Specifications

61.91
Gray
2950°C
5.22g/mL
Titanium nitride sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
Hot Pressed Material
1 each
MFCD00049596
25583-20-4
99.5%, (metals basis)

Safety and Handling

RTECSNumber : XR2230000

TSCA : TSCA