Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium nitride, Aerosol Refractory Paint, ≥99%

Chemical Identifiers

Chemical Name or Material Titanium nitride
CAS 25583-20-4
Name Note Aerosol Refractory Paint
MDL Number MFCD00049596
UN Number UN1950
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA403900CL

  SDS
alfa aesar™
403900CL
5 x 13 oz. CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA40390CLC

  SDS
alfa aesar™
40390CLC
13 oz. CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Titanium nitride
Name Note Aerosol Refractory Paint
CAS 25583-20-4
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H220-H319-H336
Extremely flammable gas.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 2.1; Packaging Group: ; Proper Shipping Name: AEROSOLS
TSCA TSCA
Assay ≥99%
MDL Number MFCD00049596
UN Number UN1950
Odor Alcohol-like
Packaging Aerosol Spray Can
Color Brown-black
Quantity 5 x 13 oz.
Safety and Handling

Danger

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H220-H319-H336
Extremely flammable gas.
Causes serious eye irritation.
May cause drowsiness or dizziness.