missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Titanium nitride, 99.7% (metal basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1451018

 View more versions of this product

Catalog No. AA1451018



Specifications

Specifications

Titanium nitride
99.7% (metal basis)
MFCD00049596
NRTOMJZYCJJWKI-UHFFFAOYSA-N
azanylidynetitanium
93091
≪10 Micron Powder
50g
25583-20-4
NTi
titanium nitride, titanium nitride tin, nitridotitanium, unii-6rw464feff, titannitrid, titaniumylidyneamine, titanium mononitride, titanium nitride, <3 mum, 6rw464feff, titanium nitride tin grade a
N#[Ti]
61.874
61.91
5.22

Chemical Identifiers

25583-20-4
61.874
NRTOMJZYCJJWKI-UHFFFAOYSA-N
93091
N#[Ti]
NTi
MFCD00049596
titanium nitride, titanium nitride tin, nitridotitanium, unii-6rw464feff, titannitrid, titaniumylidyneamine, titanium mononitride, titanium nitride, <3 mum, 6rw464feff, titanium nitride tin grade a
azanylidynetitanium
Safety and Handling

Safety and Handling

EINECSNumber : 25583-20-4

Documents