missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Titanium nitride, 99.7% (metal basis), Alfa Aesar™

$101.26 - $983.61

Chemical Identifiers

CAS 25583-20-4
Molecular Formula NTi
Molecular Weight (g/mol) 61.874
MDL Number MFCD00049596
InChI Key NRTOMJZYCJJWKI-UHFFFAOYSA-N
Synonym 6rw464feff, nitridotitanium, titanium mononitride, titanium nitride, titanium nitride tin, titanium nitride tin grade a, titanium nitride, <3 mum, titaniumylidyneamine, titannitrid, unii-6rw464feff
PubChem CID 93091
IUPAC Name azanylidynetitanium
SMILES N#[Ti]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1451018
View Documents
Thermo Fisher Scientific
1451018
50g Each for $101.26
 
AA1451030
View Documents
Thermo Fisher Scientific
1451030
250g Each for $324.65
 
AA14510A1
View Documents
Thermo Fisher Scientific
14510A1
1kg Each for $983.61
 
Specifications

Chemical Identifiers

25583-20-4
61.874
NRTOMJZYCJJWKI-UHFFFAOYSA-N
93091
N#[Ti]
NTi
MFCD00049596
6rw464feff, nitridotitanium, titanium mononitride, titanium nitride, titanium nitride tin, titanium nitride tin grade a, titanium nitride, <3 mum, titaniumylidyneamine, titannitrid, unii-6rw464feff
azanylidynetitanium

Specifications

25583-20-4
NTi
6rw464feff, nitridotitanium, titanium mononitride, titanium nitride, titanium nitride tin, titanium nitride tin grade a, titanium nitride, <3 mum, titaniumylidyneamine, titannitrid, unii-6rw464feff
NRTOMJZYCJJWKI-UHFFFAOYSA-N
azanylidynetitanium
93091
≪10 Micron Powder
2950°C
Titanium nitride
99.7% (metal basis)
MFCD00049596
Insoluble in water.
N#[Ti]
61.874
61.91
5.22
50g
Safety and Handling

Safety and Handling

EINECSNumber : 247-117-5

RTECSNumber : XR2230000

TSCA : Yes

Recommended Storage : Ambient temperatures