missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Titanium nitride, 99.7% (metal basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 14510A1

 View more versions of this product

Catalog No. AA14510A1



Specifications

Specifications

Titanium nitride
99.7% (metal basis)
MFCD00049596
Insoluble in water.
N#[Ti]
61.874
61.91
5.22
1kg
25583-20-4
NTi
6rw464feff, nitridotitanium, titanium mononitride, titanium nitride, titanium nitride tin, titanium nitride tin grade a, titanium nitride, <3 mum, titaniumylidyneamine, titannitrid, unii-6rw464feff
NRTOMJZYCJJWKI-UHFFFAOYSA-N
azanylidynetitanium
93091
≪10 Micron Powder
2950°C

Chemical Identifiers

25583-20-4
61.874
NRTOMJZYCJJWKI-UHFFFAOYSA-N
93091
N#[Ti]
NTi
MFCD00049596
6rw464feff, nitridotitanium, titanium mononitride, titanium nitride, titanium nitride tin, titanium nitride tin grade a, titanium nitride, <3 mum, titaniumylidyneamine, titannitrid, unii-6rw464feff
azanylidynetitanium
Safety and Handling

Safety and Handling

EINECSNumber : 247-117-5

RTECSNumber : XR2230000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents