missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Titanium(IV) Sulfide, 99.8% (Metal Basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1282606

 View more versions of this product

Catalog No. AA1282606



Specifications

Specifications

Titanium(IV) Sulfide
99.8%, metal basis
MFCD00049598
CFJRPNFOLVDFMJ-UHFFFAOYSA-N
bis(sulfanylidene)titanium
61544
-200 Mesh Powder
5g
12039-13-3
S2Ti
titanium sulfide tis2, titanium disulfide, titanium iv sulfide, bis sulfanylidene titanium, titanium disulphide, titansulfid, titanium iv sulfide, titanium sulfide, tis2, titanium iv sulfide-ti, titanium disulfide, crystal
S=[Ti]=S
111.987
112.03
Moisture sensitive

Chemical Identifiers

12039-13-3
111.987
CFJRPNFOLVDFMJ-UHFFFAOYSA-N
61544
S=[Ti]=S
S2Ti
MFCD00049598
titanium sulfide tis2, titanium disulfide, titanium iv sulfide, bis sulfanylidene titanium, titanium disulphide, titansulfid, titanium iv sulfide, titanium sulfide, tis2, titanium iv sulfide-ti, titanium disulfide, crystal
bis(sulfanylidene)titanium
Safety and Handling

Safety and Handling

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.

Contact with acids liberates toxic gas. Irritating to eyes, respiratory system and skin.

DOTInformation : DOT Class: 4.3

EINECSNumber : 234-883-0

Documents