Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium(IV) oxide, rutile, single crystal

Chemical Identifiers

Chemical Name or Material Titanium(IV) oxide, rutile
CAS 1317-80-2
Name Note Single crystal, 0.5mm thick x 10mm x 10mm
Formula Weight 79.9
MDL Number MFCD00011269
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA36425KT

  SDS
alfa aesar™
36425KT
1 each CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Titanium(IV) oxide, rutile
Name Note Single crystal, 0.5mm thick x 10mm x 10mm
CAS 1317-80-2
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H351
Suspected of causing cancer.
Health Hazard 3 GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Formula Weight 79.9
MDL Number MFCD00011269
Density 4.23g/mL
Odor Odorless
Quantity 1 each
Safety and Handling

Warning

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H351
Suspected of causing cancer.