missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium(IV) oxide, rutile, 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 1463109

 View more versions of this product

Catalog No. AA1463109



Specifications

Specifications

Titanium(IV) oxide
1317-80-2
O2Ti
14,9472
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
dioxotitanium
26042
79.9
99.99%
Odorless
White
10g
Rutile
(metals basis)
MFCD00011269
titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
O=[Ti]=O
79.865
CHEBI:32234
Powder
4.23g/mL
2500°C to 3000°C
1830°C to 1850°C

Chemical Identifiers

1317-80-2
79.865
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
26042
dioxotitanium
O2Ti
MFCD00011269
titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance
CHEBI:32234
O=[Ti]=O
Safety and Handling

Safety and Handling

GHS H Statement
H351
Suspected of causing cancer.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : XR2275000

TSCA : TSCA

SDS Documents