missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium(IV) oxide, rutile, 99.99% (metals basis)

$125.04 - $1,687.55

Chemical Identifiers

CAS 1317-80-2
Molecular Formula O2Ti
Molecular Weight (g/mol) 79.865
MDL Number MFCD00011269
InChI Key GWEVSGVZZGPLCZ-UHFFFAOYSA-N
Synonym anatase, brookite, flamenco, hombitan, rutile, titafrance, titania, titanium dioxide, titanium iv oxide, titanium white
PubChem CID 26042
ChEBI CHEBI:32234
IUPAC Name dioxotitanium
SMILES O=[Ti]=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1463109
View Documents
Alfa Aesar™
1463109
10g Each for $125.04
 
AA1463118
View Documents
Alfa Aesar™
1463118
50g Each for $458.05
 
AA1463130
View Documents
Alfa Aesar™
1463130
250g Each for $1,687.55
 
Specifications

Chemical Identifiers

1317-80-2
79.865
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
26042
dioxotitanium
O2Ti
MFCD00011269
anatase, brookite, flamenco, hombitan, rutile, titafrance, titania, titanium dioxide, titanium iv oxide, titanium white
CHEBI:32234
O=[Ti]=O

Specifications

1317-80-2
MFCD00011269
anatase, brookite, flamenco, hombitan, rutile, titafrance, titania, titanium dioxide, titanium iv oxide, titanium white
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
dioxotitanium
26042
79.9
99.99%
Odorless
White
10g
(metals basis)
O2Ti
14,9472
Soluble in hot concentrated sulfuric acid,in hydrofluoric acid and alkali. Insoluble in water.
O=[Ti]=O
79.865
CHEBI:32234
White powder
4.23 g/mL
2500°C to 3000°C
1830°C to 1850°C
Titanium(IV) oxide, Rutile
Safety and Handling

Safety and Handling

GHS H Statement
H351
Suspected of causing cancer.

P201-P202-P281-P308+P313-P501c

H351

EINECSNumber : 215-282-2

RTECSNumber : XR2275000

TSCA : Yes

Recommended Storage : Ambient temperatures