missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Titanium(IV) oxide, rutile, 99.9% (metals basis), Alfa Aesar™

$98.07 - $246.31

Chemical Identifiers

CAS 1317-80-2
Molecular Formula O2Ti
Molecular Weight (g/mol) 79.865
MDL Number MFCD00011269
InChI Key GWEVSGVZZGPLCZ-UHFFFAOYSA-N
Synonym anatase
PubChem CID 26042
ChEBI CHEBI:32234
IUPAC Name dioxotitanium
SMILES O=[Ti]=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4437514 View Documents Thermo Fisher Scientific
4437514
25g Each for $98.07
 
AA4437522 View Documents Thermo Fisher Scientific
4437522
100g Each for $246.31
 
Specifications

Chemical Identifiers

1317-80-2
79.865
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
26042
dioxotitanium
O2Ti
MFCD00011269
anatase
CHEBI:32234
O=[Ti]=O

Specifications

1317-80-2
O2Ti
anatase
O=[Ti]=O
79.865
CHEBI:32234
3 to 6mm Sintered Pieces
25g
99.9% (metals basis)
MFCD00011269
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
dioxotitanium
26042
79.9
4.23g/mL
Titanium(IV) oxide
Safety and Handling

Safety and Handling

EINECSNumber : 215-282-2