missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Titanium(IV) oxide, rutile, 99.9% (metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 4437522

 View more versions of this product

Catalog No. AA4437522



Specifications

Specifications

Titanium(IV) oxide
99.9% (metals basis)
MFCD00011269
Soluble in hot concentrated sulfuric acid,in hydrofluoric acid and alkali. Insoluble in water.
O=[Ti]=O
79.865
CHEBI:32234
3 to 6mm Sintered Pieces
2500°C to 3000°C
100g
1317-80-2
O2Ti
anatase
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
dioxotitanium
26042
79.9
4.23
1830°C to 1850°C

Chemical Identifiers

1317-80-2
79.865
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
26042
dioxotitanium
O2Ti
MFCD00011269
anatase
CHEBI:32234
O=[Ti]=O
Safety and Handling

Safety and Handling

P201-P202-P281-P308+P313-P501c

H351

EINECSNumber : 215-282-2

RTECSNumber : XR2275000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents