missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium(IV) oxide bis(2,4-pentanedionate)

Manufacturer:  Alfa Aesar™ 7713030

 View more versions of this product

Catalog No. AA7713030



Specifications

Specifications

Titanium (IV) oxide bis (2,4-pentanedionate)
C10H16O5Ti
bis acetylacetonato titanium iv oxide, bis 4-hydroxypent-3-en-2-one ; titanium ii oxide
CC(=CC(=O)C)O.CC(=CC(=O)C)O.O=[Ti]
264.1
262.12
184°C (decomposition)
14024-64-7
MFCD00013505
ADVORQMAWLEPOI-UHFFFAOYSA-N
4-hydroxypent-3-en-2-one;oxotitanium
74765597
Powder
250g

Chemical Identifiers

14024-64-7
264.1
ADVORQMAWLEPOI-UHFFFAOYSA-N
74765597
CC(=CC(=O)C)O.CC(=CC(=O)C)O.O=[Ti]
C10H16O5Ti
MFCD00013505
bis acetylacetonato titanium iv oxide, bis 4-hydroxypent-3-en-2-one ; titanium ii oxide
4-hydroxypent-3-en-2-one;oxotitanium
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : XR2330000

TSCA : TSCA

SDS Documents