Please login to your online account to display your discounted pricing

Titanium(IV) oxide, anatase, 99.9% (metals basis), Alfa Aesar™

$85.27 - $717.22

Chemical Identifiers

Chemical Name or Material Titanium(IV) oxide
CAS 1317-70-0
Physical Form 32Nm APS Powder, S.A. 45m&superscript_2;/g
Assay Percent Range 99.9% (metals basis)
Molecular Formula TiO2
Formula Weight 79.9
MDL Number MFCD00011269
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3995314

  SDS
alfa aesar
3995314
25g Each for $85.27

AA3995336

  SDS
alfa aesar
3995336
500g Each for $717.22

AA3995322

  SDS
alfa aesar
3995322
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form 32Nm APS Powder, S.A. 45m&superscript_2;/g
Formula Weight 79.9
Quantity 25g
Chemical Name or Material Titanium(IV) oxide
Assay Percent Range 99.9% (metals basis)
Molecular Formula TiO2
MDL Number MFCD00011269
CAS 1317-70-0
EINECS Number 215-280-1
Density 3.9g/mL