Please login to your online account to display your discounted pricing

Titanium(IV) oxide, anatase, 99.6% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Titanium(IV) oxide
CAS 1317-70-0
Name Note anatase
Molecular Formula TiO2
Formula Weight 79.90g/mol
MDL Number MFCD00011269
Merck Index 14,9472
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA3620014

 
alfa aesar
36200-14
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3620022

 
alfa aesar
36200-22
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Density 3.9
Odor Odorless
Quantity 25g
Chemical Name or Material Titanium(IV) oxide
Name Note anatase
CAS 1317-70-0
Physical Form 3 to 6mm Sintered Pieces
Assay 99.6%
Assay Percent Notes (metals basis)
Molecular Formula TiO2
Formula Weight 79.90g/mol
MDL Number MFCD00011269
Merck Index 14,9472
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H351;
Suspected of causing cancer.
Health Hazard 3 GHS P Statement
P281-P201-P202-P308+P313-P405-P501a;
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number BV6550000
TSCA TSCA
Safety and Handling

Warning

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a;
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H351;
Suspected of causing cancer.