Learn More
Titanium(IV) oxide, anatase, 99.6% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03619914
Description
Titanium(IV) oxide is used as UV-resistant cloth, skin protecting cream, face washing milk, cream milk, powder make-up. It is also used as photocatalyst, self-cleaning glass, self-cleaning ceramics, antibacterial material, air purification, sewage handling, chemical industry. It is widely utilized in cosmetics, sunscreen cream, natural white moisture protection cream, beauty and whitening cream, dawn and night cream, moistening refresher, vanishing cream, chemical fiber, plastics, printing ink coating. It is likewise utilized for improving the impressionability and opacity of the paper and used for producing titanium, ferrotitanium alloy, carbide alloy etc. in the metallurgical industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Titanium(IV) oxide | |
1317-70-0 | |
25 g | |
O2Ti | |
MFCD00011269,MFCD00210650 | |
titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance | |
GWEVSGVZZGPLCZ-UHFFFAOYSA-N | |
dioxotitanium | |
26042 | |
79.9 | |
Odorless |
Anatase | |
Powder | |
(metals basis) | |
TiO2 | |
14,9472 | |
Insoluble in water,dilute acids,organic acids,and dilute alkalis. | |
O=[Ti]=O | |
79.87 | |
CHEBI:32234 | |
99.6% | |
3.9 g/mL |
Chemical Identifiers
1317-70-0 | |
79.87 | |
GWEVSGVZZGPLCZ-UHFFFAOYSA-N | |
26042 | |
dioxotitanium |
O2Ti | |
MFCD00011269,MFCD00210650 | |
titanium dioxide, titania, titanium iv oxide, rutile, anatase, brookite, titanium white, flamenco, hombitan, titafrance | |
CHEBI:32234 | |
O=[Ti]=O |
Safety and Handling
GHS H Statement
H351
Suspected of causing cancer.
GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 215-280-1
RTECSNumber : BV6550000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only