missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Titanium(IV) oxide, catalyst support, Median pore diameter 140Å, Alfa Aesar™

Anatase, Typical properties = S.A. > 150m2/g, median pore diameter 140Å, total pore volume 0.38cc/g, packing density 0.90kg/L, contains 1% S (as SO4)

Manufacturer:  Thermo Fisher Scientific 4442930

 View more versions of this product

Catalog No. AA4442930



Specifications

Specifications

Titanium(IV) oxide
1317-70-0
MFCD00011269
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
dioxotitanium
26042
79.9
250g
1/8 in. Pellets
O2Ti
anatase
O=[Ti]=O
79.865
CHEBI:32234
0.125 in. Pellets

Chemical Identifiers

1317-70-0
79.865
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
26042
dioxotitanium
O2Ti
MFCD00011269
anatase
CHEBI:32234
O=[Ti]=O
Safety and Handling

Safety and Handling

EINECSNumber : 215-280-1

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents