missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Titanium(IV) oxide, Aeroxide(R) P25, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC384290010

 View more versions of this product

Catalog No. AC384290010



Description

Description

Specifications

Specifications

Titanium(IV) oxide
O2Ti
MFCD00011269
anatase, brookite, flamenco, hombitan, rutile, titafrance, titania, titanium dioxide, titanium iv oxide, titanium white
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
dioxotitanium
26042
79.88
≥99.5% (based on ignited material) typically
2.0% max. (1000°C, 2 hrs) (based on dried material) typically
Plastic drum
Solubility in water: insoluble. Other solubilities: soluble in hot concentrated h2so4 and hf
35 to 65 m2/g (BET)
White to Yellow
3.5 to 4.5 (4% dispersion)
13463-67-7
TiO2
15, 9628
Solubility in water: insoluble. Other solubilities: soluble in hot concentrated h2so4 and hf
O=[Ti]=O
79.88
CHEBI:32234
Powder
4.2300g/mL
1.5% max. (105°C, 2 hrs) typically
Sieve residue on 45 Micron sieve: <0.05%
4.23
2900.0°C
1855.0°C
1kg

Chemical Identifiers

13463-67-7
79.88
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
26042
dioxotitanium
O2Ti
MFCD00011269
anatase, brookite, flamenco, hombitan, rutile, titafrance, titania, titanium dioxide, titanium iv oxide, titanium white
CHEBI:32234
O=[Ti]=O
Safety and Handling

Safety and Handling

EINECSNumber : 236-675-5

SDS Documents
Promotions

Promotions