missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Titanium(IV) oxide, 99.995% (metals basis), Alfa Aesar™, Puratronic™

Manufacturer:  Thermo Fisher Scientific 1089730

 View more versions of this product

Catalog No. AA1089730



Specifications

Specifications

Titanium(IV) oxide
99.995% (metals basis)
MFCD00011269
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
dioxotitanium
26042
79.9
4.26g/mL
13463-67-7
O2Ti
anatase
O=[Ti]=O
79.865
CHEBI:32234
-22 Mesh Powder
250g

Chemical Identifiers

13463-67-7
79.865
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
26042
dioxotitanium
O2Ti
MFCD00011269
anatase
CHEBI:32234
O=[Ti]=O
Safety and Handling

Safety and Handling

EINECSNumber : 236-675-5

Documents