Please login to your online account to display your discounted pricing

Titanium(IV) oxide, 99.995% (metals basis), Alfa Aesar™, Puratronic™

$109.92 - $1,383.11

Chemical Identifiers

Chemical Name or Material Titanium(IV) oxide
CAS 13463-67-7
Physical Form -22 Mesh Powder
Assay Percent Range 99.995% (metals basis)
Molecular Formula TiO2, predominantly rutile and anatase
Formula Weight 79.9
MDL Number MFCD00011269
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1089730

 
alfa aesar
1089730
250g Each for $1,383.11

AA1089709

 
alfa aesar
1089709
10g Each for $109.92

AA1089718

 
alfa aesar
1089718
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 79.9
Quantity 250g
EINECS Number 236-675-5
Physical Form -22 Mesh Powder
Density 4.26g/mL
MDL Number MFCD00011269
Chemical Name or Material Titanium(IV) oxide
Assay Percent Range 99.995% (metals basis)
Molecular Formula TiO2, predominantly rutile and anatase
CAS 13463-67-7