missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Titanium(IV) oxide, 98+%, anatase powder, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 213581000

 View more versions of this product

Catalog No. AC213581000



Specifications

Specifications

Titanium(IV) oxide
13463-67-7
O2Ti
MFCD00011269
anatase, brookite, flamenco, hombitan, rutile, titafrance, titania, titanium dioxide, titanium iv oxide, titanium white
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
dioxotitanium
26042
79.88
98+%
Residue on 44μm sieve
0.05% max. (residue on 44 micron sieve)
4.23
White
7.3 to 8.3 (aq. suspension)
99%
98% min
TiO2
15, 9628
Solubility in water: insoluble. Other solubilities: soluble in hot concentrated h2so4 and hf
O=[Ti]=O
79.88
CHEBI:32234
Powder
4.2300g/mL
Glass bottle
Solubility in water: insoluble. Other solubilities: soluble in hot concentrated h2so4 and hf
2900.0°C
1855.0°C
100g

Chemical Identifiers

13463-67-7
79.88
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
26042
dioxotitanium
O2Ti
MFCD00011269
anatase, brookite, flamenco, hombitan, rutile, titafrance, titania, titanium dioxide, titanium iv oxide, titanium white
CHEBI:32234
O=[Ti]=O
Safety and Handling

Safety and Handling

EINECSNumber : 236-675-5

RTECSNumber : XR2275000

TSCA : TSCA

SDS Documents