missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium(IV) oxide, 20-35% in H2O colloidal dispersion

UV protection in clear coatings

Manufacturer:  Alfa Aesar™ 44517A7

 View more versions of this product

Catalog No. AA44517A7



Specifications

Specifications

Titanium (IV) oxide
13463-67-7
MFCD00011269
Insoluble in water.
O=[Ti]=O
79.865
CHEBI:32234
20 to 35% in H2O
368°C
Colloidal dispersion
O2Ti
anatase
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
dioxotitanium
26042
79.9
1.26 g/mL
5kg

Chemical Identifiers

13463-67-7
79.865
GWEVSGVZZGPLCZ-UHFFFAOYSA-N
26042
dioxotitanium
O2Ti
MFCD00011269
anatase
CHEBI:32234
O=[Ti]=O
Safety and Handling

Safety and Handling

GHS H Statement
H351
Suspected of causing cancer.

P201-P202-P281-P308+P313-P501c

H351

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents