Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium(IV) n-propoxide, VERTEC™ NPT, 98+%

Chemical Identifiers

Chemical Name or Material Titanium (IV) n-propoxide, VERTEC™ NPT
CAS 3087-37-4
Molecular Formula Ti(OCH2CH2CH3)4
Formula Weight 284.25
MDL Number MFCD00009365
UN Number UN2413
Synonym Tetra-n-propoxytitanium(IV); Tetra-n-propyl orthotitanate(IV)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4467314

 
alfa aesar™
4467314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4467336

 
alfa aesar™
4467336
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 284.25
Health Hazard 3 GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
F IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Titanium (IV) n-propoxide, VERTEC™ NPT
TSCA TSCA
Physical Form Liquid
Refractive Index 1.4986
Assay ≥98%
Molecular Formula Ti(OCH2CH2CH3)4
MDL Number MFCD00009365
CAS 3087-37-4
UN Number UN2413
Synonym Tetra-n-propoxytitanium(IV); Tetra-n-propyl orthotitanate(IV)
Boiling Point 58°C (1mmHg)
Melting Point 20°C
Quantity 25g
Flash Point 42°C (109°F)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H319
Flammable liquid and vapor.
Causes serious eye irritation.
Density 0.955g/mL
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: TETRAPROPYLORTHOTITANATE
Safety and Handling

Warning

GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
F IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H319
Flammable liquid and vapor.
Causes serious eye irritation.