Learn More
Titanium(IV) n-butoxide, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 223192500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acidSpecifications
Titanium(IV) n-butoxide | |
5593-70-4 | |
Colorless to Yellow | |
310.0°C to 314.0°C | |
Authentic | |
Glass bottle | |
1.486 | |
MFCD00009433 | |
0.99 | |
Solubility in water: rapidly hydrolized by water. Other solubilities: miscible with most common organic solvents | |
CCCCO.CCCCO.CCCCO.CCCCO.[Ti] | |
340.35 | |
50-80 mPa.s (20°C) | |
98+% |
99% | |
-75°C | |
0.9900g/mL | |
47°C | |
98+% | |
C16H36O4Ti | |
Ti[O(CH2)3CH3]4 | |
250 g | |
butyl titanate, butan-1-ol; titanium, titanium n-butoxide, tetrabutyl orthotitanate, tetrabutyl titanate, 1-butanol; titanium, tetra butyl orthotitanate, tetrabutyl ortho-titanate | |
FPCJKVGGYOAWIZ-UHFFFAOYSA-N | |
butan-1-ol;titanium | |
25203927 | |
340.35 | |
Liquid |
Chemical Identifiers
5593-70-4 | |
340.35 | |
FPCJKVGGYOAWIZ-UHFFFAOYSA-N | |
25203927 | |
CCCCO.CCCCO.CCCCO.CCCCO.[Ti] |
C16H36O4Ti | |
MFCD00009433 | |
butyl titanate, butan-1-ol; titanium, titanium n-butoxide, tetrabutyl orthotitanate, tetrabutyl titanate, 1-butanol; titanium, tetra butyl orthotitanate, tetrabutyl ortho-titanate | |
butan-1-ol;titanium |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
May cause drowsiness or dizziness.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If skin irritation occurs: Get medical advice/at
GHS Signal Word: Danger
EINECSNumber : 227-006-8
RTECSNumber : XR1585000
TSCA : TSCA
Recommended Storage : the product darkens with age
RUO – Research Use Only