missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium(IV) n-butoxide, 99+%

Manufacturer:  Alfa Aesar™ 7712409

 View more versions of this product

Catalog No. AA7712409



Specifications

Specifications

Titanium (IV) n-butoxide
C16H40O4Ti
MFCD00009433
4148236
Soluble in organic solvents; Decomposes in water
CCCCO.CCCCO.CCCCO.CCCCO.[Ti]
344.359
340.36
≥99%
Alcohol-like
Moisture Sensitive
Yellow
10g
5593-70-4
Ti[O(CH2)3CH3]4
UN2924
butyl titanate, butan-1-ol; titanium, titanium n-butoxide, tetrabutyl orthotitanate, tetrabutyl titanate, 1-butanol; titanium, tetra butyl orthotitanate, tetrabutyl ortho-titanate
FPCJKVGGYOAWIZ-UHFFFAOYSA-N
butan-1-ol;titanium
25203927
Liquid
0.998g/mL
1.4900
310°C to 314°C
-55°C
41°C (170°F)

Chemical Identifiers

5593-70-4
344.359
FPCJKVGGYOAWIZ-UHFFFAOYSA-N
25203927
CCCCO.CCCCO.CCCCO.CCCCO.[Ti]
C16H40O4Ti
MFCD00009433
butyl titanate, butan-1-ol; titanium, titanium n-butoxide, tetrabutyl orthotitanate, tetrabutyl titanate, 1-butanol; titanium, tetra butyl orthotitanate, tetrabutyl ortho-titanate
butan-1-ol;titanium
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

RTECSNumber : XR1585000

TSCA : TSCA

SDS Documents