Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium(IV) methylphenoxide, tech.

$22.64 - $157.00

Chemical Identifiers

Chemical Name or Material Titanium (IV) methylphenoxide
CAS 28503-70-0
Purity Grade Technical Grade
Molecular Formula C28H28O4Ti
Formula Weight 476.43
MDL Number MFCD00061534
Synonym Titanium(IV) cresylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA8945409

 
alfa aesar™
8945409
10g Each for $22.64

AA8945430

 
alfa aesar™
8945430
250g Each for $72.80

AA89454A1

 
alfa aesar™
89454A1
1kg Each for $157.00
Description & Specifications

Specifications

Formula Weight 476.43
Chemical Name or Material Titanium (IV) methylphenoxide
TSCA TSCA
Purity Grade Technical Grade
Physical Form Liquid
Sensitivity Moisture Sensitive
Molecular Formula C28H28O4Ti
MDL Number MFCD00061534
CAS 28503-70-0
Synonym Titanium(IV) cresylate
Boiling Point 230° to 240°C (0.01mmHg)
Color Dark red
Melting Point −36°C
Quantity 10g
Flash Point 66°C (150°F)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 1.055g/mL
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.