missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium(IV) methylphenoxide, tech.

Manufacturer:  Alfa Aesar™ 89454A1

 View more versions of this product

Catalog No. AA89454A1



Specifications

Specifications

Titanium (IV) methylphenoxide
Technical Grade
MFCD00061534
476.43
1.055g/mL
230°C to 240°C (0.01mmHg)
-36°C
66°C (150°F)
28503-70-0
C28H28O4Ti
Titanium(IV) cresylate
Liquid
Moisture Sensitive
Red
1kg

Chemical Identifiers

28503-70-0
C28H28O4Ti
MFCD00061534
Titanium(IV) cresylate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents