Learn More
Titanium(IV) methoxide, 95%, Thermo Scientific Chemicals
$146.95 - $1869.81
Chemical Identifiers
CAS | 992-92-7 |
---|---|
Molecular Formula | Ti(OCH3)4·(CH3OH)x, x<1 |
MDL Number | MFCD00014883 |
Synonym | tetramethyl titanate, titanium tetramethoxide, titanium iv methoxide, methanolate; titanium 4+, titanium tetra methanolate, methanol, titanium 4+ salt, titanium methoxide, methanol, titanium 4+ salt 4:1, titanium 4+ tetrakis methoxide, titanium 4+ ion tetrakis methoxide |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA7713714
|
Thermo Scientific Chemicals
07713714 |
25 g |
Each for $146.95
|
|
|||||
AA7713722
|
Thermo Scientific Chemicals
07713722 |
100 g |
Each for $484.15
|
|
|||||
AA7713736
|
Thermo Scientific Chemicals
07713736 |
500 g |
Each for $1,869.81
|
|
|||||
Description
Titanium(IV) methoxide is used for the preparation of polyoxotitanates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
992-92-7 | |
Ti(OCH3)4·(CH3OH)x, x<1 | |
MFCD00014883 | |
tetramethyl titanate, titanium tetramethoxide, titanium iv methoxide, methanolate; titanium 4+, titanium tetra methanolate, methanol, titanium 4+ salt, titanium methoxide, methanol, titanium 4+ salt 4:1, titanium 4+ tetrakis methoxide, titanium 4+ ion tetrakis methoxide |
Specifications
>243°C (52mmHg) | |
992-92-7 | |
25 g | |
MFCD00014883 | |
48°C (118°F) | |
Slightly soluble in organic solvents. | |
Moisture sensitive |
200°C to 210°C | |
Granular/Lump | |
Ti(OCH3)4·(CH3OH)x, x<1 | |
UN3181 | |
tetramethyl titanate, titanium tetramethoxide, titanium iv methoxide, methanolate; titanium 4+, titanium tetra methanolate, methanol, titanium 4+ salt, titanium methoxide, methanol, titanium 4+ salt 4:1, titanium 4+ tetrakis methoxide, titanium 4+ ion tetrakis methoxide | |
95% | |
Titanium (IV) methoxide |
Safety and Handling
GHS H Statement
H228-H302-H312-H332-H319
Flammable solid.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P370+P378q-P501c
H228-H302+H312-H315-H318-H335
DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
EINECSNumber : 213-596-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only