missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium(IV) methoxide, 95%

Manufacturer:  Alfa Aesar™ 7713722

 View more versions of this product

Catalog No. AA7713722



Specifications

Specifications

Titanium (IV) methoxide
Ti(OCH3)4·(CH3OH)x, x<1
UN3181
Granular/Lump
Moisture Sensitive
200°C to 210°C
48°C (118°F)
992-92-7
MFCD00014883
methanol, titanium 4+ salt
95%
>243°C (52mmHg)
100g

Chemical Identifiers

992-92-7
Ti(OCH3)4·(CH3OH)x, x<1
MFCD00014883
methanol, titanium 4+ salt
Safety and Handling

Safety and Handling

GHS H Statement
H228-H302-H312-H332-H319
Flammable solid.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement
P210-P261-P280-P305+P351+P338-P304+P340-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.

TSCA : TSCA

Documents