missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Titanium(IV) methoxide, 95%

Manufacturer:  Alfa Aesar™ 7713714

 View more versions of this product

Catalog No. AA7713714



Specifications

Specifications

Titanium (IV) methoxide
Ti(OCH3)4·(CH3OH)x, x<1
UN3181
Slightly soluble in organic solvents.
95%
>243°C (52mmHg)
25g
992-92-7
MFCD00014883
methanol, titanium 4+ salt, methanol, titanium 4+ salt 4:1, methanolate; titanium 4+, tetramethyl titanate, titanium 4+ ion tetrakis methoxide, titanium 4+ tetrakis methoxide, titanium iv methoxide, titanium methoxide, titanium tetra methanolate, titanium tetramethoxide
Granular/ lump
Moisture sensitive
200°C to 210°C
48°C (118°F)

Chemical Identifiers

992-92-7
Ti(OCH3)4·(CH3OH)x, x<1
MFCD00014883
methanol, titanium 4+ salt, methanol, titanium 4+ salt 4:1, methanolate; titanium 4+, tetramethyl titanate, titanium 4+ ion tetrakis methoxide, titanium 4+ tetrakis methoxide, titanium iv methoxide, titanium methoxide, titanium tetra methanolate, titanium tetramethoxide
Safety and Handling

Safety and Handling

GHS H Statement
H228-H302-H312-H332-H319
Flammable solid.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.

P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P370+P378q-P501c

H228-H302+H312-H315-H318-H335

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.

EINECSNumber : 213-596-4

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents