missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Titanium(IV) isopropoxide, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 194700050

 View more versions of this product

Catalog No. AC194700050



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
Specifications

Specifications

Titanium(IV) isopropoxide
>15.0°C
240.0°C
Authentic
Glass bottle
1.4654 to 1.4684
5 mL
11,92; 12,90; 13,13; 14,61; 15,308; 16,54; 17,347
15,9636
Solubility in water: hydrolysis. Other solubilities: soluble in most common organic solvents
CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-].[Ti+4]
284.26
4.3 mPa.s (25°C)
98+%
546-68-9
0.9500g/mL
46°C
16.6 to 17.3% (Ti)
C12H28O4Ti
Ti[OCH(CH3)2]4
01,II,382
0.95
titanium tetraisopropanolate, titanium iv isopropoxide, tetraisopropyl orthotitanate, titanium isopropoxide, titanium tetraisopropylate, titanium isopropylate, ti isopropylate, tetraisopropoxytitanium iv, isopropyl orthotitanate, tetraisopropyl titanate
VXUYXOFXAQZZMF-UHFFFAOYSA-N
propan-2-olate;titanium(4+)
11026
284.26
Liquid

Chemical Identifiers

546-68-9
284.26
titanium tetraisopropanolate, titanium iv isopropoxide, tetraisopropyl orthotitanate, titanium isopropoxide, titanium tetraisopropylate, titanium isopropylate, ti isopropylate, tetraisopropoxytitanium iv, isopropyl orthotitanate, tetraisopropyl titanate
propan-2-olate;titanium(4+)
C12H28O4Ti
VXUYXOFXAQZZMF-UHFFFAOYSA-N
11026
CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-].[Ti+4]
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if inhaled.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remov

GHS Signal Word: Danger

EINECSNumber : 208-909-6

Recommended Storage : Product may darken during storage

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only