Learn More
Titanium(IV) isopropoxide, 97+%, Thermo Scientific Chemicals
Catalyzes the regio selective ring opening of 2,3 epoxy alcohols and acids
Supplier: Thermo Scientific Chemicals 07711536
Description
Catalyzes the regio selective ring opening of 2,3 epoxy alcohols and acids.Titanium(IV) isopropoxide is used as a precursor for the preparation of titanium and barium-strontium-titanate thin films. It is useful to make porous titanosilicates and potential ion-exchange materials for cleanup of radioactive wastes. It is an active component of sharpless epoxidation as well as involved in the synthesis of chiral epoxides. In Kulinkovich reaction, it is involved as a catalyst in the preparation of cyclopropanes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
232°C | |
16°C to 20°C | |
Colorless to Yellow | |
500 g | |
Ti[OCH(CH3)2]4 | |
UN2413 | |
46°C (115°F) | |
titanium tetraisopropanolate, titanium iv isopropoxide, tetraisopropyl orthotitanate, titanium isopropoxide, titanium tetraisopropylate, titanium isopropylate, ti isopropylate, tetraisopropoxytitanium iv, isopropyl orthotitanate, tetraisopropyl titanate | |
VXUYXOFXAQZZMF-UHFFFAOYSA-N | |
propan-2-olate;titanium(4+) | |
11026 | |
≥97% | |
Moisture sensitive |
Titanium (IV) isopropoxide | |
546-68-9 | |
Liquid | |
C12H28O4Ti | |
MFCD00008871 | |
3679474 | |
14,9480 | |
Soluble in anhydrous ethanol, ether, benzene and chloroform; Decomposes in water | |
CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-].[Ti+4] | |
284.219 | |
284.23 | |
1.464 | |
0.955 g/mL |
Chemical Identifiers
546-68-9 | |
284.219 | |
VXUYXOFXAQZZMF-UHFFFAOYSA-N | |
11026 | |
CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-].[Ti+4] |
C12H28O4Ti | |
MFCD00008871 | |
titanium tetraisopropanolate, titanium iv isopropoxide, tetraisopropyl orthotitanate, titanium isopropoxide, titanium tetraisopropylate, titanium isopropylate, ti isopropylate, tetraisopropoxytitanium iv, isopropyl orthotitanate, tetraisopropyl titanate | |
propan-2-olate;titanium(4+) |
Safety and Handling
GHS H Statement
H226-H319
Flammable liquid and vapor.
Causes serious eye irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P311-P370+P378q-P501c
H226-H319-H331-H335-H336
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: TETRAPROPYLORTHOTITANATE
EINECSNumber : 208-909-6
RTECSNumber : NT8060000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only